Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD8B
Entrez Gene ID
101103712
Gene Name
CD8b molecule
Gene Aliases
-
Chromosome Location
Chr.3: 62710718 - 62729416 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004007264.4 XP_004007313.2 5 - 6 666 64

View Full Product Details