Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HEYL
Entrez Gene ID
101119343
Gene Name
hes related family bHLH transcription factor with YRPW motif like
Gene Aliases
-
Chromosome Location
Chr.1: 14807636 - 14822939 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004001829.4 XP_004001878.2 1 - 2 114 96

View Full Product Details