Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BCL7C
Entrez Gene ID
101103285
Gene Name
BAF chromatin remodeling complex subunit BCL7C
Gene Aliases
-
Chromosome Location
Chr.24: 27569731 - 27574433 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004020916.4 XP_004020965.2 5 - 6 664 68

View Full Product Details