Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS51
Entrez Gene ID
101120711
Gene Name
VPS51 subunit of GARP complex
Gene Aliases
-
Chromosome Location
Chr.21: 45105777 - 45119198 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027959926.1 XP_027815727.1 1 - 2 278 106

View Full Product Details