Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXL3
Entrez Gene ID
100169937
Gene Name
F-box and leucine rich repeat protein 3
Gene Aliases
-
Chromosome Location
Chr.10: 56319818 - 56343133 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129739.1 NP_001123211.1 3 - 4 646 55
XM_027973501.1 XP_027829302.1 4 - 5 994 55
XM_027973502.1 XP_027829303.1 4 - 5 723 55
GenBank mRNA EF643523.1 - 3 - 4 646 55
EF643524.1 - 3 - 4 502 55

View Full Product Details