Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPINK4
Entrez Gene ID
101104631
Gene Name
serine peptidase inhibitor Kazal type 4
Gene Aliases
-
Chromosome Location
Chr.2: 39928893 - 39936397 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004004147.4 XP_004004196.1 1 - 2 117 72

View Full Product Details