Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRAP2
Entrez Gene ID
101117592
Gene Name
melanocortin 2 receptor accessory protein 2
Gene Aliases
-
Chromosome Location
Chr.8: 58360313 - 58387241 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027972719.1 XP_027828520.1 2 - 3 320 101

View Full Product Details