Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SH3BP4
Entrez Gene ID
101104281
Gene Name
SH3 domain binding protein 4
Gene Aliases
-
Chromosome Location
Chr.1: 6000347 - 6099196 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027967359.1 XP_027823160.1 4 - 5 2819 68

View Full Product Details