Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRTCAP2
Entrez Gene ID
101116525
Gene Name
keratinocyte associated protein 2
Gene Aliases
-
Chromosome Location
Chr.1: 111521531 - 111524158 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004002576.4 XP_004002625.2 2 - 3 259 85

View Full Product Details