Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NHS
Entrez Gene ID
101107499
Gene Name
NHS actin remodeling regulator
Gene Aliases
-
Chromosome Location
Chr.X: 16680475 - 16741008 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015104813.2 XP_014960299.2 4 - 5 576 65

View Full Product Details