Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC25A47
Entrez Gene ID
101119167
Gene Name
solute carrier family 25 member 47
Gene Aliases
-
Chromosome Location
Chr.18: 65729564 - 65737400 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027957399.1 XP_027813200.1 3 - 4 674 76
XM_027957400.1 XP_027813201.1 2 - 3 1692 76

View Full Product Details