Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NPHS2
Entrez Gene ID
101121618
Gene Name
NPHS2 stomatin family member, podocin
Gene Aliases
-
Chromosome Location
Chr.12: 65089835 - 65108814 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027976124.1 XP_027831925.1 1 - 2 310 82

View Full Product Details