Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASB4
Entrez Gene ID
101117496
Gene Name
ankyrin repeat and SOCS box containing 4
Gene Aliases
-
Chromosome Location
Chr.4: 14570701 - 14656886 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004007737.4 XP_004007786.2 1 - 2 239 100

View Full Product Details