Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC58
Entrez Gene ID
101108095
Gene Name
leucine rich repeat containing 58
Gene Aliases
-
Chromosome Location
Chr.1: 203096799 - 203115404 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004002953.4 XP_004003002.2 2 - 3 723 80

View Full Product Details