Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HESX1
Entrez Gene ID
101103514
Gene Name
HESX homeobox 1
Gene Aliases
-
Chromosome Location
Chr.19: 45862607 - 45885186 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012099865.3 XP_011955255.1 3 - 4 1516 96
XM_015102359.2 XP_014957845.1 1 - 2 6647 96

View Full Product Details