Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMX1A
Entrez Gene ID
101111592
Gene Name
LIM homeobox transcription factor 1 alpha
Gene Aliases
-
Chromosome Location
Chr.1: 124618547 - 124790272 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027957471.1 XP_027813272.1 8 - 9 1054 98

View Full Product Details