Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EHD4
Entrez Gene ID
101115625
Gene Name
EH domain containing 4
Gene Aliases
-
Chromosome Location
Chr.7: 37296810 - 37382918 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004010473.4 XP_004010522.2 1 - 2 323 69

View Full Product Details