Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL17F
Entrez Gene ID
101104186
Gene Name
interleukin 17F
Gene Aliases
-
Chromosome Location
Chr.20: 26221446 - 26228928 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004018888.4 XP_004018937.2 2 - 3 429 79

View Full Product Details