Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NIP7
Entrez Gene ID
101120941
Gene Name
nucleolar pre-rRNA processing protein NIP7
Gene Aliases
-
Chromosome Location
Chr.14: 39254856 - 39286617 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004015098.4 XP_004015147.1 1 - 2 270 66
XM_015100516.2 XP_014956002.1 1 - 2 274 66

View Full Product Details