Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DMRT2
Entrez Gene ID
101110302
Gene Name
doublesex and mab-3 related transcription factor 2
Gene Aliases
-
Chromosome Location
Chr.2: 74575045 - 74581952 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004004343.3 XP_004004392.1 2 - 3 942 101
XM_015093191.2 XP_014948677.1 2 - 3 942 101
XM_027964525.1 XP_027820326.1 2 - 3 942 101

View Full Product Details