Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CBX5
Entrez Gene ID
100307036
Gene Name
chromobox 5
Gene Aliases
-
Chromosome Location
Chr.3: 141809051 - 141843378 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166190.1 NP_001159662.1 2 - 3 320 80
XM_012158957.2 XP_012014347.1 3 - 4 536 80
XM_012158964.3 XP_012014354.1 3 - 4 449 80
XM_012158975.3 XP_012014365.1 3 - 4 467 80
GenBank mRNA GQ221053.1 - 2 - 3 320 80

View Full Product Details