The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_004014974.4 | XP_004015023.1 | 1 - 2 | 266 | 82 |
XM_012189737.3 | XP_012045127.1 | 1 - 2 | 268 | 82 |