Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARFIP2
Entrez Gene ID
100302307
Gene Name
ADP ribosylation factor interacting protein 2
Gene Aliases
-
Chromosome Location
Chr.15: 49751302 - 49756334 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162549.1 NP_001156021.1 6 - 7 871 87
XM_012095242.3 XP_011950632.1 7 - 8 1003 87
XM_012095243.1 XP_011950633.1 4 - 5 616 87
XM_027978975.1 XP_027834776.1 7 - 8 971 87
GenBank mRNA FJ969418.1 - 6 - 7 871 87

View Full Product Details