Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMO1
Entrez Gene ID
101102757
Gene Name
LIM domain only 1
Gene Aliases
-
Chromosome Location
Chr.15: 47433861 - 47475531 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012095820.3 XP_011951210.1 3 - 4 420 89
XM_015101044.2 XP_014956530.1 3 - 4 504 89

View Full Product Details