Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
KISS1
Entrez Gene ID
101107719
Gene Name
KiSS-1 metastasis suppressor
Gene Aliases
-
Chromosome Location
Chr.12: 1561901 - 1568776 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306104.1 NP_001293033.1 2 - 3 358 77
XM_027975561.1 XP_027831362.1 2 - 3 1223 77
GenBank mRNA JQ716394.1 - 2 - 3 358 77
KY356393.1 - 1 - 2 489 77

View Full Product Details