Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ST3GAL5
Entrez Gene ID
101104977
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 5
Gene Aliases
-
Chromosome Location
Chr.3: 61756671 - 61819198 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306114.1 NP_001293043.1 1 - 2 124 125
XM_012169456.3 XP_012024846.2 2 - 3 125
XM_027965734.1 XP_027821535.1 2 - 3 125
GenBank mRNA KF055859.1 - 1 - 2 124 125

View Full Product Details