Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TIMP3
Entrez Gene ID
100217407
Gene Name
TIMP metallopeptidase inhibitor 3
Gene Aliases
-
Chromosome Location
Chr.3: 189199882 - 189254687 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166187.1 NP_001159659.1 3 - 4 313 76
GenBank mRNA FJ384972.1 - 1 - 2 85 76
GQ221039.1 - 3 - 4 313 76

View Full Product Details