Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
ANGPTL4
Entrez Gene ID
101104317
Gene Name
angiopoietin like 4
Gene Aliases
-
Chromosome Location
Chr.5: 15327030 - 15333559 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308589.1 NP_001295518.1 2 - 3 688 121
GenBank mRNA KF873613.1 - 2 - 3 688 121

View Full Product Details