Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VAMP7
Entrez Gene ID
100307050
Gene Name
vesicle associated membrane protein 7
Gene Aliases
-
Chromosome Location
Chr.X: 87059480 - 87145684 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166204.1 NP_001159676.1 6 - 7 586 96
XM_012106120.3 XP_011961510.1 7 - 8 729 96
GenBank mRNA GQ221059.1 - 6 - 7 586 96

View Full Product Details