Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
OAZ2
Entrez Gene ID
101107870
Gene Name
ornithine decarboxylase antizyme 2
Gene Aliases
-
Chromosome Location
Chr.7: 46443609 - 46456016 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290006.2 NP_001276935.1 2 - 2 274 110
GenBank mRNA GT644620.1 - 2 - 2 298 110
GT873426.1 - 2 - 2 193 110

View Full Product Details