Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BRS3
Entrez Gene ID
443044
Gene Name
bombesin receptor subtype 3
Gene Aliases
BRS-3
Chromosome Location
Chr.X: 107329955 - 107334425 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009215.1 NP_001009215.1 2 - 3 927 84
GenBank mRNA AF108210.1 - 2 - 3 927 84

View Full Product Details