Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCNG1
Entrez Gene ID
101119213
Gene Name
cyclin G1
Gene Aliases
-
Chromosome Location
Chr.5: 78371944 - 78379193 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287473.1 NP_001274402.1 1 - 2 273 89
XM_004009046.4 XP_004009095.1 2 - 3 485 89
XM_012177580.3 XP_012032970.1 2 - 3 481 89
GenBank mRNA KC539855.1 - 1 - 2 273 89

View Full Product Details