Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCNG1
Entrez Gene ID
101119213
Gene Name
cyclin G1
Gene Aliases
-
Chromosome Location
Chr.5: 78371944 - 78379193 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287473.1 NP_001274402.1 4 - 5 703 78
XM_004009046.4 XP_004009095.1 5 - 6 915 78
XM_012177580.3 XP_012032970.1 5 - 6 911 78
GenBank mRNA KC539855.1 - 4 - 5 703 78

View Full Product Details