Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCNG1
Entrez Gene ID
101119213
Gene Name
cyclin G1
Gene Aliases
-
Chromosome Location
Chr.5: 78371944 - 78379193 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287473.1 NP_001274402.1 5 - 6 894 71
XM_004009046.4 XP_004009095.1 6 - 7 1106 71
GenBank mRNA KC539855.1 - 5 - 6 894 71

View Full Product Details