Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CGA
Entrez Gene ID
443538
Gene Name
glycoprotein hormones, alpha polypeptide
Gene Aliases
-
Chromosome Location
Chr.8: 55695479 - 55713877 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009464.1 NP_001009464.1 2 - 3 171 99
GenBank mRNA X16977.1 - - 171 99

View Full Product Details