Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARF4
Entrez Gene ID
100302304
Gene Name
ADP ribosylation factor 4
Gene Aliases
-
Chromosome Location
Chr.19: 45599810 - 45617281 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162546.1 NP_001156018.1 1 - 2 62 99
GenBank mRNA FJ969400.1 - 1 - 2 62 99

View Full Product Details