Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LYPLA1
Entrez Gene ID
101107971
Gene Name
lysophospholipase 1
Gene Aliases
-
Chromosome Location
Chr.9: 37727421 - 37745107 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306103.1 NP_001293032.1 1 - 1 659 80
GenBank mRNA KJ000742.1 - 1 - 1 659 80

Gene Symbol
LOC114115289
Entrez Gene ID
114115289
Gene Name
acyl-protein thioesterase 1
Gene Aliases
-
Chromosome Location
Chr.6: 40443462 - 40445879 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027970878.1 XP_027826679.1 1 - 1 655 80
XM_027970879.1 XP_027826680.1 2 - 2 553 80

View Full Product Details