Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC39A2
Entrez Gene ID
100302553
Gene Name
solute carrier family 39 member 2
Gene Aliases
-
Chromosome Location
Chr.5: 44161756 - 44164647 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163601.1 NP_001157073.1 2 - 3 244 94
XM_012180583.2 XP_012035973.1 2 - 3 846 94
GenBank mRNA FJ937951.1 - 2 - 3 244 94

View Full Product Details