Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL22
Entrez Gene ID
100885770
Gene Name
interleukin 22
Gene Aliases
IL-22
Chromosome Location
Chr.3: 162601245 - 162606507 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260478.1 NP_001247407.1 4 - 5 476 91
GenBank mRNA HE617662.1 - 4 - 5 476 91

View Full Product Details