Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDYN
Entrez Gene ID
101105430
Gene Name
prodynorphin
Gene Aliases
-
Chromosome Location
Chr.13: 55460659 - 55462736 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001280677.1 NP_001267606.1 1 - 2 133 51
GenBank mRNA JQ514279.1 - 1 - 2 133 51

View Full Product Details