Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
B4GALT1
Entrez Gene ID
100620048
Gene Name
beta-1,4-galactosyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.2: 40023816 - 40074220 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001246211.1 NP_001233140.1 1 - 2 446 85
GenBank mRNA JF510320.1 - 1 - 2 446 85

View Full Product Details