Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FTO
Entrez Gene ID
100125353
Gene Name
FTO alpha-ketoglutarate dependent dioxygenase
Gene Aliases
-
Chromosome Location
Chr.14: 23529695 - 23956080 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104931.1 NP_001098401.1 2 - 3 129 87
XM_027977360.1 XP_027833161.1 2 - 3 226 87
GenBank mRNA EU072419.1 - 2 - 3 129 87

View Full Product Details