Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
GRP
Entrez Gene ID
443332
Gene Name
gastrin releasing peptide
Gene Aliases
-
Chromosome Location
Chr.23: 64287896 - 64300976 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009321.1 NP_001009321.1 2 - 3 473 83
GenBank mRNA S75723.1 - 2 - 3 473 83

View Full Product Details