Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OAZ3
Entrez Gene ID
101119005
Gene Name
ornithine decarboxylase antizyme 3
Gene Aliases
-
Chromosome Location
Chr.1: 107442707 - 107449407 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290359.1 NP_001277288.1 4 - 5 593 90
NM_001301418.1 NP_001288347.1 4 - 5 625 90

View Full Product Details