Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
ASMT
Entrez Gene ID
101114754
Gene Name
acetylserotonin O-methyltransferase
Gene Aliases
HIOMT
Chromosome Location
Chr.X: 604548 - 626110 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306120.1 NP_001293049.1 - 347 88
GenBank mRNA JQ647512.1 - - 270 88
KC290950.1 - - 347 88

View Full Product Details