Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSPAN9
Entrez Gene ID
100180629
Gene Name
tetraspanin 9
Gene Aliases
NET5, TNE5, TSPAN5, Tspan-9
Chromosome Location
Chr.3: 227378891 - 227569370 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134305.1 NP_001127777.1 6 - 7 642 100
XM_027965719.1 XP_027821520.1 8 - 9 804 100
XM_027965720.1 XP_027821521.1 7 - 8 828 100
GenBank mRNA EU797610.1 - 6 - 7 642 100

View Full Product Details