Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AVPR1A
Entrez Gene ID
100528010
Gene Name
arginine vasopressin receptor 1A
Gene Aliases
V1R, V1aR
Chromosome Location
Chr.3: 167691716 - 167694581 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199792.1 NP_001186721.1 1 - 2 980 96
GenBank mRNA HM856595.1 - 1 - 2 980 96

View Full Product Details