Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP2E1
Entrez Gene ID
100568285
Gene Name
cytochrome P450, family 2, subfamily E, polypeptide 1
Gene Aliases
-
Chromosome Location
Chr.22: 55607721 - 55618200 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001245972.1 NP_001232901.1 5 - 6 855 80
GenBank mRNA HQ263378.1 - 5 - 6 855 80

View Full Product Details