Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
B4GALT7
Entrez Gene ID
100316848
Gene Name
beta-1,4-galactosyltransferase 7
Gene Aliases
-
Chromosome Location
Chr.5: 39357533 - 39365819 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168634.1 NP_001162105.1 5 - 6 950 67
GenBank mRNA FN568103.1 - 5 - 6 950 67

View Full Product Details