Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCML1
Entrez Gene ID
101103129
Gene Name
Scm polycomb group protein like 1
Gene Aliases
-
Chromosome Location
Chr.X: 16742353 - 16759651 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012106304.3 XP_011961694.1 7 - 8 1322 62
XM_012106306.3 XP_011961696.1 6 - 7 1241 62
XM_012106307.3 XP_011961697.1 6 - 7 1238 62
XM_012106308.3 XP_011961698.1 7 - 8 818 62

View Full Product Details